ENA429360681 Enamine

1-(3-amino-2-methylphenyl)-3,3-dimethylurea

  • MDL number MFCD21809469
  • Cas Number : 1525599-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190451

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190451
Availability details for
Quantity
  • Availability
  • Add to Cart