ENA429360682 Enamine

2-hexanamidoacetic acid

  • MDL number MFCD09909949
  • Cas Number : 24003-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190453

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190453
Availability details for
Quantity
  • Availability
  • Add to Cart