ENA429360684 Enamine

tert-butyl 3-(3-aminophenyl)propanoate

  • MDL number MFCD23972938
  • Cas Number : 269071-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190455
Availability details for
Quantity
  • Availability
  • Add to Cart