ENA429360685 Enamine

benzyl 2-sulfanylacetate

  • MDL number MFCD04070643
  • Cas Number : 7383-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190456

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190456
Availability details for
Quantity
  • Availability
  • Add to Cart