ENA429360689 Enamine

8-methylpyrido[2,3-b]pyrazine

  • MDL number MFCD09881189
  • Cas Number : 1023817-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189529

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189529
Availability details for
Quantity
  • Availability
  • Add to Cart