ENA429360692 Enamine

1-[5-(3-bromophenyl)-1,2,4-oxadiazol-3-yl]methanamine hydrochloride

  • MDL number MFCD28118843
  • Cas Number : 1803580-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189537
Availability details for
Quantity
  • Availability
  • Add to Cart