ENA429360693 Enamine

6-azaspiro[3.5]nonane

  • MDL number MFCD14581274
  • Cas Number : 71873-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190475
Availability details for
Quantity
  • Availability
  • Add to Cart