ENA429360694 Enamine

2-phenylpropane-1-thiol

  • MDL number MFCD23790708
  • Cas Number : 34366-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-190476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190476
Availability details for
Quantity
  • Availability
  • Add to Cart