ENA429360696 Enamine

3,5-dimethyl-4-phenyl-1,2-oxazole

  • MDL number MFCD16301498
  • Cas Number : 4345-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190484
Availability details for
Quantity
  • Availability
  • Add to Cart