ENA429360702 Enamine

8-(piperidin-1-yl)quinolin-5-amine

  • MDL number MFCD12412244
  • Cas Number : 856090-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190502
Availability details for
Quantity
  • Availability
  • Add to Cart