ENA429360704 Enamine

2-fluoro-4-(trifluoromethyl)phenol

  • MDL number MFCD00061285
  • Cas Number : 77227-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190504

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-190504
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart