ENA429360708 Enamine

4-(1-methyl-1H-pyrazol-4-yl)-4-oxobutanoic acid

  • MDL number MFCD10694891
  • Cas Number : 1152553-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190524

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190524
Availability details for
Quantity
  • Availability
  • Add to Cart