ENA429360710 Enamine

ethyl 2-(piperidin-4-yloxy)acetate hydrochloride

  • MDL number MFCD12546300
  • Cas Number : 209112-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190527

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190527
Availability details for
Quantity
  • Availability
  • Add to Cart