ENA429360718 Enamine

1-hydroxy-1,3-dihydro-2,1-benzoxaborole-5-carboxylic acid

  • MDL number MFCD27992243
  • Cas Number : 1801711-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189558

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189558
Availability details for
Quantity
  • Availability
  • Add to Cart