ENA429360719 Enamine

methyl 2-bromo-3-(4-methylphenyl)propanoate

  • MDL number MFCD11559349
  • Cas Number : 7141-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190567
Availability details for
Quantity
  • Availability
  • Add to Cart