ENA429360722 Enamine

1-[3-(3-bromophenyl)-1,2,4-oxadiazol-5-yl]methanamine hydrochloride

  • MDL number MFCD28118845
  • Cas Number : 1803566-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189559
Availability details for
Quantity
  • Availability
  • Add to Cart