ENA429360723 Enamine

5-methoxy-1-phenylpentan-2-amine

  • MDL number MFCD16822036
  • Cas Number : 1249980-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190579

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190579
Availability details for
Quantity
  • Availability
  • Add to Cart