ENA429360724 Enamine

ethyl 5-(chlorosulfonyl)-1-benzothiophene-2-carboxylate

  • MDL number MFCD24231455
  • Cas Number : 1547967-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190581

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190581
Availability details for
Quantity
  • Availability
  • Add to Cart