ENA429360725 Enamine

3-(4-fluorophenyl)cyclopentan-1-amine

  • MDL number MFCD19347595
  • Cas Number : 1267444-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190586

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190586
Availability details for
Quantity
  • Availability
  • Add to Cart