ENA429360739 Enamine

3-ethyl-3-methyl-2,3,4,5-tetrahydro-1H-1,4-benzodiazepine-2,5-dione

  • MDL number MFCD23794446
  • Cas Number : 1431141-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190638
Availability details for
Quantity
  • Availability
  • Add to Cart