ENA429360740 Enamine

2-amino-1-phenylpentan-1-ol

  • MDL number MFCD20696096
  • Cas Number : 773794-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-72
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190639
Availability details for
Quantity
  • Availability
  • Add to Cart