ENA429360743 Enamine

5-(difluoromethyl)-3-ethyl-1H-1,2,4-triazole

  • MDL number MFCD14610984
  • Cas Number : 1247413-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190658
Availability details for
Quantity
  • Availability
  • Add to Cart