ENA429360745 Enamine

(3,5-diethyl-1-phenyl-1H-pyrazol-4-yl)methanamine

  • MDL number MFCD23273150
  • Cas Number : 1506103-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189564

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189564
Availability details for
Quantity
  • Availability
  • Add to Cart