ENA429360751 Enamine

[5-(methylsulfanyl)thiophen-2-yl]methanol

  • MDL number MFCD06202671
  • Cas Number : 773868-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189594
Availability details for
Quantity
  • Availability
  • Add to Cart