ENA429360753 Enamine

5-(trifluoromethyl)-1,2-benzoxazol-3-amine

  • MDL number MFCD11847114
  • Cas Number : 868271-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190692

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190692
Availability details for
Quantity
  • Availability
  • Add to Cart