ENA429360755 Enamine

2-cyclobutoxy-4,6-dimethylpyridine-3-carboxylic acid

  • MDL number MFCD16716552
  • Cas Number : 1247523-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190700

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190700
Availability details for
Quantity
  • Availability
  • Add to Cart