ENA429360773 Enamine

4-(1H-1,2,3-benzotriazol-1-yl)pyrimidin-2-amine

  • MDL number MFCD23992098
  • Cas Number : 1542059-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190724
Availability details for
Quantity
  • Availability
  • Add to Cart