ENA429360775 Enamine

tert-butyl 2-(tert-butoxy)acetate

  • MDL number MFCD26054310
  • Cas Number : 55666-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189599
Availability details for
Quantity
  • Availability
  • Add to Cart