ENA429360776 Enamine

2-(3-chlorophenyl)-2-(piperazin-1-yl)acetonitrile

  • MDL number MFCD09817086
  • Cas Number : 1017388-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190746

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190746
Availability details for
Quantity
  • Availability
  • Add to Cart