ENA429360777 Enamine

tert-butyl N-tert-butyl-N-(2-oxoethyl)carbamate

  • MDL number MFCD24469702
  • Cas Number : 156591-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190749

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190749
Availability details for
Quantity
  • Availability
  • Add to Cart