ENA429360778 Enamine

2-amino-N,3-dimethyl-N-({5H,6H,7H-pyrrolo[2,1-c][1,2,4]triazol-3-yl}methyl)butanamide

  • MDL number MFCD12885232
  • Cas Number : 1218462-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190750
Availability details for
Quantity
  • Availability
  • Add to Cart