ENA429360783 Enamine

4-{5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazin-7-yl}pyrimidin-2-amine

  • MDL number MFCD23990647
  • Cas Number : 1501212-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190764
Availability details for
Quantity
  • Availability
  • Add to Cart