ENA429360784 Enamine

8-fluoro-2,3,4,5-tetrahydro-1,4-benzoxazepin-5-one

  • MDL number MFCD22416207
  • Cas Number : 844648-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190767

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190767
Availability details for
Quantity
  • Availability
  • Add to Cart