ENA429360789 Enamine

4-(chloromethyl)-2-methylpyrimidine

  • MDL number MFCD10697077
  • Cas Number : 89967-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190776
Availability details for
Quantity
  • Availability
  • Add to Cart