ENA429360798 Enamine

2-bromo-3-fluoro-4-(trifluoromethyl)pyridine

  • MDL number MFCD16609968
  • Cas Number : 1227577-05-0
  • Molecular Weight : 243.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-190801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190801
Availability details for
Quantity
  • Availability
  • Add to Cart