ENA429360800 Enamine

1-[4-(3-aminocyclobutyl)piperazin-1-yl]ethan-1-one

  • MDL number MFCD20703237
  • Cas Number : 1272668-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190804

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190804
Availability details for
Quantity
  • Availability
  • Add to Cart