ENA429360803 Enamine

methyl 5-ethyl-3-phenyl-1,2-oxazole-4-carboxylate

  • MDL number MFCD22370413
  • Cas Number : 929890-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190835

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190835
Availability details for
Quantity
  • Availability
  • Add to Cart