ENA429360806 Enamine

3-(2-chlorophenyl)-5-ethyl-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD19092941
  • Cas Number : 91961-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190838

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190838
Availability details for
Quantity
  • Availability
  • Add to Cart