ENA429360807 Enamine

tert-butyl 2-cyano-2-methylacetate

  • MDL number MFCD12545840
  • Cas Number : 66476-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190845

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190845
Availability details for
Quantity
  • Availability
  • Add to Cart