ENA429360808 Enamine

4-(pyridin-4-ylmethyl)piperidin-4-ol dihydrochloride

  • MDL number MFCD28010457
  • Cas Number : 1021205-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190847

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190847
Availability details for
Quantity
  • Availability
  • Add to Cart