ENA429360810 Enamine

3-[(6-nitroquinazolin-4-yl)amino]propane-1,2-diol

  • MDL number MFCD23199318
  • Cas Number : 1478560-05-2
  • Molecular Weight : 264.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-190849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190849
Availability details for
Quantity
  • Availability
  • Add to Cart