ENA429360820 Enamine

2-chloro-1-cyclopentylethan-1-one

  • MDL number MFCD09955061
  • Cas Number : 932-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190882
Availability details for
Quantity
  • Availability
  • Add to Cart