ENA429360823 Enamine

2-(2-methylpropyl)pyrimidin-5-amine

  • MDL number MFCD10697097
  • Cas Number : 944901-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190887

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190887
Availability details for
Quantity
  • Availability
  • Add to Cart