ENA429360826 Enamine

ethyl 1-methyl-4-oxocyclohexane-1-carboxylate

  • MDL number MFCD17018639
  • Cas Number : 147905-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190899

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190899
Availability details for
Quantity
  • Availability
  • Add to Cart