ENA429360827 Enamine

methyl 4-(sulfamoylamino)benzoate

  • MDL number MFCD26245634
  • Cas Number : 127903-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190904

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190904
Availability details for
Quantity
  • Availability
  • Add to Cart