ENA429360828 Enamine

potassium trifluoro[2-fluoro-5-(trifluoromethyl)phenyl]boranuide

  • MDL number MFCD18861580

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190908
Availability details for
Quantity
  • Availability
  • Add to Cart