ENA429360834 Enamine

methyl 6-chloro-1-methyl-1H-1,3-benzodiazole-2-carboxylate

  • MDL number MFCD26058458
  • Cas Number : 1565595-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190938
Availability details for
Quantity
  • Availability
  • Add to Cart