ENA429360843 Enamine

3-(1H-pyrazol-1-yl)pyridine-2-carbonitrile

  • MDL number MFCD24132061
  • Cas Number : 1504980-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190977
Availability details for
Quantity
  • Availability
  • Add to Cart