ENA429360847 Enamine

thiolan-3-ylmethanamine

  • MDL number MFCD19667166
  • Cas Number : 933701-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189637

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189637
Availability details for
Quantity
  • Availability
  • Add to Cart