ENA429360849 Enamine

tert-butyl (2S)-2-aminohexanoate

  • MDL number MFCD20690671
  • Cas Number : 15911-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191007
Availability details for
Quantity
  • Availability
  • Add to Cart